LIF432342366 Life Chemicals Inc.

methyl (piperidin-4-ylsulfonyl)acetate hydrochloride

  • MDL number MFCD24842732
  • Cas Number : 1448125-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart