LIF432342371 Life Chemicals Inc.

tert-butyl (1,4-dihydroxybutan-2-yl)carbamate

  • MDL number MFCD23378730
  • Cas Number : 156627-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart