LIF432342372 Life Chemicals Inc.

4'-(trifluoromethyl)-[1,1'-biphenyl]-3-sulfonyl chloride

  • MDL number MFCD01631887
  • Cas Number : 731779-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart