LIF432342374 Life Chemicals Inc.

4-(phenylsulfanyl)piperidine hydrochloride

  • MDL number MFCD03840144
  • Cas Number : 101798-66-7
  • Molecular Weight : 229.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart