LIF432342375 Life Chemicals Inc.

3-[4-(trifluoromethyl)phenyl]aniline

  • MDL number MFCD03424671
  • Cas Number : 400747-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart