LIF432342377 Life Chemicals Inc.

2-cyclopentylethan-1-amine hydrochloride

  • MDL number MFCD09866300
  • Cas Number : 684221-26-9
  • Molecular Weight : 149.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart