LIF432342383 Life Chemicals Inc.

2-cyclohexylacetonitrile

  • MDL number MFCD00058667
  • Cas Number : 4435-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart