LIF432342385 Life Chemicals Inc.

4-(4-chlorobenzyl)piperidine hydrochloride

  • MDL number MFCD03840138
  • Cas Number : 36968-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart