LIF443508475 Life Chemicals Inc.

1,3-oxazinan-2-imine hydrochloride

  • MDL number MFCD28165869
  • Cas Number : 173556-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart