LIF444783848 Life Chemicals Inc.

3-(chloromethyl)-5-methyl-1H-pyrazole hydrochloride

  • MDL number MFCD28023517
  • Cas Number : 278798-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart