LIF444783849 Life Chemicals Inc.

3-(chloromethyl)-4,5,6,7-tetrahydro-1H-indazole hydrochloride

  • MDL number MFCD28388516
  • Cas Number : 114538-05-5
  • Molecular Weight : 207.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(chloromethyl)-4,5,6,7-tetrahydro-1H-indazole hydrochloride
Inchi Code 1S/C8H11ClN2.ClH/c9-5-8-6-3-1-2-4-7(6)10-11-8;/h1-5H2,(H,10,11);1H
Inchi Key ROEHQMDMNFLJQZ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart