LIF444783852 Life Chemicals Inc.

2-(piperidin-3-yl)-5-(trifluoromethyl)-1,3,4-oxadiazole hydrochloride

  • MDL number MFCD28388518
  • Cas Number : 1820712-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart