LIF444783853 Life Chemicals Inc.

3-amino-1,2,3,4-tetrahydronaphthalen-2-ol hydrochloride

  • MDL number MFCD28388520
  • Cas Number : 103028-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart