LIF444783854 Life Chemicals Inc.

N-(2,2,2-trifluoroethyl)piperazine-1-sulfonamide hydrochloride

  • MDL number MFCD28369755
  • Cas Number : 1820613-25-9
  • Molecular Weight : 283.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart