LIF444783855 Life Chemicals Inc.

N-(2,2,2-trifluoroethyl)-1,4-diazepane-1-sulfonamide hydrochloride

  • MDL number MFCD28369756
  • Cas Number : 1820666-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart