LIF444783857 Life Chemicals Inc.

piperazine-1-sulfonamide hydrochloride

  • MDL number MFCD28388522
  • Cas Number : 1403952-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart