LIF444783858 Life Chemicals Inc.

8-bromo-5-methoxy-1,2,3,4-tetrahydro-1,6-naphthyridine

  • MDL number MFCD28369849
  • Cas Number : 1820711-91-8
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart