LIF444783859 Life Chemicals Inc.

8-bromo-N,N-dimethyl-1,2,3,4-tetrahydro-1,6-naphthyridin-5-amine

  • MDL number MFCD28369850
  • Cas Number : 1820666-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart