LIF444783860 Life Chemicals Inc.

6-methoxy-7-methyl-1,2,3,4-tetrahydro-1,5-naphthyridine

  • MDL number MFCD28388523
  • Cas Number : 1820716-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart