LIF444783862 Life Chemicals Inc.

4-methyl-1,5-naphthyridin-2-amine

  • MDL number MFCD28388524
  • Cas Number : 1820640-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart