LIF444783863 Life Chemicals Inc.

2-chloro-4-methyl-1,5-naphthyridine

  • MDL number MFCD28388525
  • Cas Number : 1820706-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart