LIF444783865 Life Chemicals Inc.

N,N,6-trimethyl-1,2,3,4-tetrahydro-1,7-naphthyridin-8-amine

  • MDL number MFCD28388528
  • Cas Number : 1820639-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart