LIF444783866 Life Chemicals Inc.

1-(5-(aminomethyl)-2-methoxyphenyl)-N,N-dimethylmethanamine dihydrochloride

  • MDL number MFCD28369866
  • Cas Number : 1820687-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart