LIF444783867 Life Chemicals Inc.

2-(5-formyl-2-methoxyphenyl)acetonitrile

  • MDL number MFCD28388531
  • Cas Number : 1820608-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart