LIF444783873 Life Chemicals Inc.

methyl 2-(2-imino-6-(methylsulfonyl)benzo[d]thiazol-3(2H)-yl)acetate hydrobromide

  • MDL number MFCD26940474
  • Cas Number : 1949836-92-3
  • Molecular Weight : 381.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart