LIF444783879 Life Chemicals Inc.

5,6-dimethyl-3-(prop-2-yn-1-yl)benzo[d]thiazol-2(3H)-imine hydrobromide

  • MDL number MFCD26940485
  • Cas Number : 1949816-32-3
  • Molecular Weight : 297.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart