LIF444783881 Life Chemicals Inc.

3-allyl-5,7-dimethylbenzo[d]thiazol-2(3H)-imine hydrobromide

  • MDL number MFCD26940490
  • Cas Number : 1949816-46-9
  • Molecular Weight : 299.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart