LIF444783915 Life Chemicals Inc.

3-allyl-6-bromobenzo[d]thiazol-2(3H)-imine hydrobromide

  • MDL number MFCD28369892
  • Cas Number : 2034154-20-4
  • Molecular Weight : 350.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart