LIF444783943 Life Chemicals Inc.

4-((4-(aminomethyl)pyridin-2-yl)oxy)cyclohexan-1-one oxalate

  • MDL number MFCD28369941
  • Cas Number : 1820650-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart