LIF444783945 Life Chemicals Inc.

4-(pyrrolidin-3-yl)phenol hydrobromide

  • MDL number MFCD28388689
  • Cas Number : 38175-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart