LIF444783948 Life Chemicals Inc.

(1R,3r,5S)-3-(pyrimidin-2-yloxy)-8-azabicyclo[3.2.1]octane dihydrochloride

  • MDL number MFCD28369993
  • Cas Number : 1820686-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart