LIF444783949 Life Chemicals Inc.

3-(pyrazin-2-yloxy)-8-azabicyclo[3.2.1]octane sesquihydrochloride

  • MDL number MFCD29046750
  • Cas Number : 1820706-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart