LIF444783954 Life Chemicals Inc.

2-(5-(2-aminoethyl)-1,2,4-oxadiazol-3-yl)acetamide hemioxalate

  • MDL number MFCD28370000
  • Cas Number : 1820614-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart