LIF444783959 Life Chemicals Inc.

3-(methylthio)-1-(3-(thiophen-2-yl)-1,2,4-oxadiazol-5-yl)propan-1-amine oxalate

  • MDL number MFCD28370007
  • Cas Number : 1820649-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart