LIF444783960 Life Chemicals Inc.

1-(3-(2-fluorophenyl)-1,2,4-oxadiazol-5-yl)-3-(methylthio)propan-1-amine oxalate

  • MDL number MFCD28370008
  • Cas Number : 1820705-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart