LIF444783961 Life Chemicals Inc.

1-(3-benzyl-1,2,4-oxadiazol-5-yl)-3-(methylthio)propan-1-amine oxalate

  • MDL number MFCD28370009
  • Cas Number : 1820703-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart