LIF444783964 Life Chemicals Inc.

1-(3-cyclopentyl-1,2,4-oxadiazol-5-yl)-N-methylmethanamine hydrochloride

  • MDL number MFCD28388798
  • Cas Number : 1807988-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart