LIF444783974 Life Chemicals Inc.

((2,3,5,6-tetramethylphenyl)sulfonyl)glycine

  • MDL number MFCD02333806
  • Cas Number : 379250-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart