LIF444783982 Life Chemicals Inc.

methyl 2-amino-4-(furan-2-yl)thiophene-3-carboxylate

  • MDL number MFCD03422510
  • Cas Number : 670271-04-2
  • Molecular Weight : 223.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart