LIF444783994 Life Chemicals Inc.

3-((3-(trifluoromethyl)phenyl)sulfonamido)propanoic acid

  • MDL number MFCD03620134
  • Cas Number : 612042-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart