LIF444784021 Life Chemicals Inc.

1-(cyclobutylmethyl)piperazine dihydrochloride

  • MDL number MFCD04596089
  • Cas Number : 877859-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart