LIF444784041 Life Chemicals Inc.

2-chloro-3-methanesulfonylpyridine

  • MDL number MFCD17016076
  • Cas Number : 70682-09-6
  • Molecular Weight : 191.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart