LIF444784042 Life Chemicals Inc.

ethyl piperazine-2-carboxylate dihydrochloride

  • MDL number MFCD22381964
  • Cas Number : 129798-91-0
  • Molecular Weight : 231.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart