LIF444784048 Life Chemicals Inc.

2-(chloromethyl)-1-ethyl-1H-imidazole hydrochloride

  • MDL number MFCD09908411
  • Cas Number : 135206-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart