LIF444784052 Life Chemicals Inc.

4-chloro-5,6,7,8-tetrahydroquinazoline

  • MDL number MFCD12755923
  • Cas Number : 1125-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart