LIF444784053 Life Chemicals Inc.

2-chloro-6-(1H-imidazol-1-yl)pyridine

  • MDL number MFCD18368361
  • Cas Number : 184097-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart