LIF444784056 Life Chemicals Inc.

2-acetamido-N-(2-benzoyl-4-bromophenyl)acetamide

  • MDL number MFCD00525060
  • Cas Number : 313502-80-6
  • Molecular Weight : 375.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart