LIF444784057 Life Chemicals Inc.

(1-acetylpiperidin-4-yl)methyl 4-methylbenzenesulfonate

  • MDL number MFCD21097900
  • Cas Number : 846057-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart