LIF444784058 Life Chemicals Inc.

(1-acetylpyrrolidin-3-yl)methyl 4-methylbenzenesulfonate

  • MDL number MFCD21097894
  • Cas Number : 1353979-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart