LIF444784074 Life Chemicals Inc.

ethyl 2-acetamido-4-phenylthiazole-5-carboxylate

  • MDL number MFCD00807206
  • Cas Number : 113019-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart